Typpipullo (Schlenk), vastaanotto (nitrogen receiver), PTFE neulaventtiilahanalla
Toimittaja: Lenz Laborglas GmbH & CO.KG
03360622 03360628 03360858 03360837 03360828 03360849 03360870
LENZ03360622EA 89.7 EUR
LENZ03360622 LENZ03360628 LENZ03360858 LENZ03360837 LENZ03360828 LENZ03360849 LENZ03360870
Typpipullo (Schlenk), vastaanotto (nitrogen receiver), PTFE neulaventtiilahanalla
Kolvit ja erlenmeyerit Typpikolvi
DURAN® borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With conical ground joint, PTFE needle-valve stopcock and inscription label.