Vacuum stopcocks
Toimittaja: Lenz Laborglas GmbH & CO.KG
02901804 02902106 02962910 02902910 02922910 02901402 02921402 02961402 02961804 02921804 02941402 02941804 02942910 02962106 02922408 02942408 02962408 02902408 02922106 02942106
189-2102EA 71.3 EUR
189-2102 189-2103 LENZ02962910 189-2105 189-2125 LENZ02901402 LENZ02921402 LENZ02961402 LENZ02961804 189-2122 189-2111 189-2112 189-2115 LENZ02962106 189-2124 189-2114 LENZ02962408 LENZ02902408 189-2123 189-2113
Vacuum stopcocks
Hanat
Vacuum stopcocks, DURAN® with side arms and hollow NS glass plug.
  • Excellent chemical resistance
  • Excellent temperature resistance
Leakage rate according to DIN 12540-1.