Sleeves for conical ground joints
Toimittaja: Lenz Laborglas GmbH & CO.KG
01101514 01101519 01101524 01101529 01101545
LENZ01101514EA 36.7 EUR
LENZ01101514 LENZ01101519 LENZ01101524 LENZ01101529 LENZ01101545
Sleeves for conical ground joints
NS-liitosholkit Liitosholkit
Disposable ground joint sleeves. thin walled, made of PTFE.
  • Sleeves have a wall thickness of 0,05 mm
  • Usable to 260 °C
  • Outstanding chemical resistance