Metal base moulds
Toimittaja: LEICA HISTOLOGY
3803084 3803082 3803080 3803083 3803081
LEIH3803084EA 157 EUR
LEIH3803084 LEIH3803082 LEIH3803080 LEIH3803083 LEIH3803081
Metal base moulds
Muotit Moulds for Histology
Metal base moulds enable tissue to be positioned and embedded in wax when using routine or biopsy cassettes. These re-usable base moulds are made from stainless steel and the internal rounded corners allow for easy removal of the formed wax block.
  • Available in packs of 10 or 12
  • Can be used on all standard size cassettes