Glove box, stainless steel, filtered, Protector®
Toimittaja: LABCONCO
5065537 5065535 5065531 5065533
LBCP5065537EA 86310 EUR
LBCP5065537 LBCP5065535 LBCP5065531 LBCP5065533
Glove box, stainless steel, filtered, Protector®
Hansikaaskaapit
These glove boxes with stainless steel liner provide inlet and outlet HEPA or ULPA filtration and a leak-tight physical barrier to protect the operator from exposure to potentially hazardous materials. Available with two or four glove ports (double version).
  • LCD display, large viewing area (double glove boxes have two viewing areas), space-saving inner and outer transfer chamber doors that pivot upward
  • Two 25-watt fluorescent lights; switches for light and electrical duplex
  • Built-in blower with speed control capable of airflow from 15 CFM to 85 CFM
  • Dry-