Glove box, fibreglass, HEPA filtered, Protector®

Toimittaja: LABCONCO
5065035 5065031
LBCP5065035EA 33870 EUR
LBCP5065035 LBCP5065031
Glove box, fibreglass, HEPA filtered, Protector®
Hansikaaskaapit
This filtered glove box with fibreglass liner provides inlet and outlet HEPA filtration and a leak-tight physical barrier to protect the operator from exposure to potentially hazardous materials. The glove box may also be appropriate for use as a containment isolator for applications that include compounds assigned ISO 4 and ISO 5 Banding Levels.

  • LCD display provides real time monitoring of volume dilution rates in cubic feet per minute (CFM), air changes/minute (ACM), and main chamber pressure (inches of water)
  • Built-in blower with speed control capable
More Product Information
Order Now

TEKNISET TIEDOT

Tyyppi Protector®
Environmentally Preferable
Liitännät 208 - 230 V, 10 amps, 50/60 Hz
Tarkasteluikkuna (L × K) 914,5×701 mm
L×S×K ulkomitat 1623×787×1161 mm
Paino 227 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR