Blower transition adapters
Toimittaja: LABCONCO
4722401 4861700
LBCP4722401EA 0 EUR
LBCP4722401 LBCP4861700
Blower transition adapters
Ducting Supplies
Epoxy-coated steel blower transition adapters fit all Labconco coated steel blowers.
Adapter allows you to connect thermoplastic duct to the exhaust side of a blower to create an exhaust stack. A nominal size PVC duct fits inside the opening.