Horizontal laminar flow, Purifier®
Toimittaja: LABCONCO
3888425 3873006 3873008 3873009 3873025 3873026 3873028 3873029 3888405 3888406 3873005 3889729 3888426 3888428 3888429 3889705 3889706 3889708 3889709 3889725 3889726 3889728 3888408
LBCP3888425EA 0 EUR
LBCP3888425 LBCP3873006 LBCP3873008 LBCP3873009 LBCP3873025 LBCP3873026 LBCP3873028 LBCP3873029 LBCP3888405 LBCP3888406 LBCP3873005 LBCP3889729 LBCP3888426 LBCP3888428 LBCP3888429 LBCP3889705 LBCP3889706 LBCP3889708 LBCP3889709 LBCP3889725 LBCP3889726 LBCP3889728 LBCP3888408
Horizontal laminar flow, Purifier®
Vetokaapit ja kotelot Laminaarivirtauskaapit
It offers a choice to protect materials that do not generate harmful aerosols or vapors. Its been designed with 99,99% efficient HEPA filter, the air from this minimizes cross contamination in the work area and provides a particulate free work environment. It has wide range of applications including plant tissue culture research, media preparation, electronic part inspection, syringe filling, medical device assembly and non hazardous parenteral drug formulation. These benches should never be used in conjunction with biohazardous material, toxins or radionuclides.
  • Differential pressure