Open access transfer station, PuriCare®
Toimittaja: LABCONCO
3820020 3820023 3820033
LBCP3820020EA 0 EUR
LBCP3820020 LBCP3820023 LBCP3820033
Open access transfer station, PuriCare®
Vetokaapit ja kotelot Laminaarivirtauskaapit
These PuriCare Open Access Transfer Station offer utmost efficiency for cage changing operations. Open access on two sides provides ample workspace for one or two operators to transfer animals from dirty cages to clean ones. This enclosure provides personnel, animal and environmental protection. HEPA-filtered vertical airflow provides Class 100 clean air in the work area to protect the animals from cross-contamination. At the same time, room air is drawn into grilles located on the work surface directly below the sash openings to ensure containment and protect the operator from allergens.More Product Information