Laboratory fume hoods, Protector®, XStream®
Toimittaja: LABCONCO
110410021
LBCP110410021EA 0 EUR
LBCP110410021
Laboratory fume hoods, Protector®, XStream®
Vetokaapit ja kotelot Vetokaappi kemikaaleille
The Protector® XStream® benchtop laboratory hoods are chemical resistant, fiberglass reinforced, composite liner and baffle. These are designed for general chemical applications.
  • Durable glacier white, dry powder-coated steel exterior with by-pass flow design
  • One-piece molded fiberglass interior and pre-set baffle with flame spread index less than 25 per ASTM E-84
  • Baffles are removable for cleaning
  • Removable front and side panels and front and interior access panels for access to plumbing and electrical wiring
  • Two tempered safety glass vertical-rising sash