Dual Position Cap for Culture Tubes
Toimittaja: LABCON
3500-800-000-9
LBCN3500-800-000-9EA 155 EUR
LBCN3500-800-000-9
Dual Position Cap for Culture Tubes
Tulpat ja korkit Koeputkitulpat
Tube caps are made of low density polyethylene for a secure seal and are available in a fluted plug style or dual position design that permits sample aspiration.
Polyethylene caps are not autoclavable, but are available sterile with many of the tube packages.