Tubings, insulated
Toimittaja: LAUDA
LZS001 LZS007 LZS009 LZS021 LZS002 LZS019 LZS020 LZS022 LZS011 LZS012 LZS023
LAUDLZS001EA 87 EUR
LAUDLZS001 LAUDLZS007 LAUDLZS009 615-1138 LAUDLZS002 LAUDLZS019 LAUDLZS020 LAUDLZS022 LAUDLZS011 LAUDLZS012 LAUDLZS023
Tubings, insulated
Letku Siirtoputki
This insulated tubing is used with thermostats.
*Silicone tubes should not used in conjunction with silicone oils.