Shaking incubators, Varioshake series
Toimittaja: LAUDA
L003052 L003053 L003054
LAUDL003052EA 8060 EUR
LAUDL003052 LAUDL003053 LAUDL003054
Shaking incubators, Varioshake series
Ravistelijat Inkubaatioravistelija
Shaking incubators with orbital movement for precise and reproducible research.
  • LED display shows temperature and enables setting in steps of 0,1 °C
  • User-friendly microprocessor technology
  • Membrane keyboard with distinctive symbols
  • PID type temperature control
  • Limit values for over and under temperature alarm (optical and acoustic) can be set
  • Timer, microprocessor-controlled, with remaining time display and acoustic signal at the end of the process
  • Timer adjustable from 1 min. up to 999 hours
  • Hinged acrylic glass hood
The models have a cooling coilMore Product Information