Heating and cooling process thermostats, Integral T
Toimittaja: LAUDA
L002663 L002664 L002665 L002666 L002667 L002668 L002669 L002670
LAUDL002663EA 0 EUR
LAUDL002663 LAUDL002664 LAUDL002665 LAUDL002666 LAUDL002667 LAUDL002668 LAUDL002669 LAUDL002670
Heating and cooling process thermostats, Integral T
Kiertotermostaatit Suljetut termostaattihaudejärjestelmät Jäähdyttävät termostaattihauteet
Integral T are process thermostats with an integrated cooling system for dynamic temperature control within external circuits.
  • PowerAdapt system uses the maximum possible amount of heat permitted by the power supply system
  • Low water level and adjustable over-temperature protection with acoustic alarm for use with flammable and non-flammable liquids
  • Extremely powerful pressure pump
  • Integrated port for external PT100, second external PT100 feasible via an interface module
  • Remote fault indication through floating contact
  • Option for upgrading up to two