Gas Sampling Balloons
Toimittaja: LAMAPLAST
80601 80602 80604 80603 80605
LAMA80601EA 43.4 EUR
LAMA80601 LAMA80602 LAMA80604 LAMA80603 LAMA80605
Gas Sampling Balloons
Näytteenottimet Kaasunäytteenottimet
Gas sampling balloons are made from natural latex.
  • Volume ranges from 1500 to 8000 ml
  • Colour: pale yellow/red
The 1500 ml balloons have a blowpipe diameter of 4,9 mm, with a blowpipe length of 95 mm. The 2000 and 4000 ml balloons have a blowpipe diameter of 5,85 mm, with a blowpipe length of 86 mm. The 5000 and 8000 ml balloons have a blowpipe diameter of 4,8 mm, with a blowpipe length of 93 mm.
Pakkaus:
Each ballon is packaged in a single plastic bag.