1,2,3-Trifluoro-5-nitrobenzene 99%

Toimittaja: Thermo Fisher Scientific
Vaara

L20302.06 L20302.14
L20302.06EA 175 EUR
L20302.06 L20302.14
1,2,3-Trifluoro-5-nitrobenzene 99%
1,2,3-Trifluoro-5-nitrobenzene

Purity: 99% CAS No.: 66684-58-0 Molecular Formula: C6H2F3NO2 Formula Weight: 177.08 Flash Point: 75°(167°F) Refractive Index: 1.4860 Beilstein Reference: 2455018 UN No.: 2810 MDL No.: MFCD00456803

Kaava: C₆H₂F₃NO₂
Tiheys: 1,52
Leimahduspiste: 75 °C (167 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00456803
CAS-numero: 66684-58-0
EINECS: 266-447-0
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR