2-(Trifluoromethyl)phenylglyoxal hydrate 98% (dry weight)

Toimittaja: Thermo Fisher Scientific
Varoitus

L20280.03 L20280.MD
L20280.03EA 314 EUR
L20280.03 L20280.MD
2-(Trifluoromethyl)phenylglyoxal hydrate 98% (dry weight)
2-(Trifluoromethyl)phenylglyoxal hydrate

Purity: 98% CAS No.: 745783-91-9 Molecular Formula: C9H5F3O2.xH2O Formula Weight: 202.13(anhy) Melting Point: 36-43° MDL No.: MFCD05664102

Kaava: C₉H₇F₃O₃
Sulamispiste: 36…43 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD06801323
CAS-numero: 745783-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR