(3-Methyl-2-butenyl)tributyltin 95%

Toimittaja: Thermo Fisher Scientific
Vaara

L20214.03 L20214.06
L20214.03EA 139 EUR
L20214.03 L20214.06
(3-Methyl-2-butenyl)tributyltin 95%
(3-Methyl-2-butenyl)tributyltin

Purity: 95% CAS No.: 53911-92-5 Molecular Formula: C17H36Sn Formula Weight: 359.19 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4880 Boiling Point: 283° Beilstein Reference: 3588742 UN No.: 2788 MDL No.: MFCD01863649

Kaava: C₁₇H₃₆Sn
Kiehumispiste: 283 °C
Tiheys: 1,042
Leimahduspiste: >110 °C (230 °F)
MDL numero: MFCD01863649
CAS-numero: 53911-92-5
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR