trans-Bis(tributylstannyl)ethylene 96%

Toimittaja: Thermo Fisher Scientific
Vaara

L20130.03 L20130.06
L20130.03EA 225 EUR
L20130.03 L20130.06
trans-Bis(tributylstannyl)ethylene 96%
trans-Bis(tributylstannyl)ethylene

Purity: 96% CAS No.: 14275-61-7 Molecular Formula: C26H56Sn2 Formula Weight: 606.15 Storage Temperature: 2°C to 8°C Refractive Index: 1.5020 Boiling Point: 185°/0.01mm Beilstein Reference: 3613803 UN No.: 2788 MDL No.: MFCD01631299

Kaava: C₂₆H₅₆Sn₂
Kiehumispiste: 185 °C (0,01 mmHg)
Tiheys: 1,140
MDL numero: MFCD01631299
CAS-numero: 14275-61-7
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR