4-Chloronicotinic acid ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

L20029.03 L20029.06
L20029.03EA 194 EUR
L20029.03 L20029.06
4-Chloronicotinic acid ≥97%
4-Chloronicotinic acid

Purity: 97% CAS No.: 10177-29-4 Molecular Formula: C6H4ClNO2 Formula Weight: 157.56 Melting Point: ca 165° dec. Beilstein Reference: 9255849

Kaava: C₆H₄ClNO₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00128860
CAS-numero: 10177-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR