Methyl-2-amino-5-fluorobenzoate 98%

Toimittaja: Thermo Fisher Scientific

L19778.06 L19778.14
L19778.06EA 138 EUR
L19778.06 L19778.14
Methyl-2-amino-5-fluorobenzoate 98%
Methyl-2-amino-5-fluorobenzoate

Purity: 98% CAS No.: 319-24-4 Molecular Formula: C8H8FNO2 Formula Weight: 169.16 Flash Point: 132°(269°F) Melting Point: 21-25° Boiling Point: 100-102°/0.5mm Beilstein Reference: 2803210 MDL No.: MFCD03791308

Kaava: C₈H₈FNO₂
Kiehumispiste: 100…102 °C (0,5 mmHg)
Sulamispiste: 21…25 °C
Tiheys: 1,260
Leimahduspiste: 132 °C (269 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03791308
CAS-numero: 319-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR