8-Kinoliinikarbaldehydi ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

L19642.03 L19642.06
L19642.03EA 340 EUR
L19642.03 L19642.06
8-Kinoliinikarbaldehydi ≥98%
8-Kinoliinikarbaldehydi

Purity: 98+% CAS No.: 38707-70-9 Molecular Formula: C10H7NO Formula Weight: 157.17 Melting Point: 92-94° Beilstein Reference: 113039

Kaava: C₁₀H₇NO
Sulamispiste: 92…94 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00805837
CAS-numero: 38707-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR