(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

L19361.03 L19361.06
L19361.03EA 73.6 EUR
L19361.03 L19361.06
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid ≥98%
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid

Purity: 98+% CAS No.: 26250-84-0 Molecular Formula: C11H19NO4 Formula Weight: 229.28 Melting Point: 121-125° Beilstein Reference: 3652038 Optical Rotation: -46° (c=1 in methanol) MDL No.: MFCD00151904

Kaava: C₁₁H₁₉NO₄
Sulamispiste: 121…125 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00151904
CAS-numero: 26250-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR