4-Methoxy-1,5-dihydro-2H-pyrrol-2-one 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

L19344.03 L19344.06
L19344.03EA 165 EUR
L19344.03 L19344.06
4-Methoxy-1,5-dihydro-2H-pyrrol-2-one 99%
4-Methoxy-1,5-dihydro-2H-pyrrol-2-one

Purity: 99% CAS No.: 69778-83-2 Molecular Formula: C5H7NO2 Formula Weight: 113.12 Melting Point: 130-134° Beilstein Reference: 1524505 MDL No.: MFCD00071564

Kaava: C₅H₇NO₂
Sulamispiste: 130…134 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00071564
CAS-numero: 69778-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR