1-Bromi-4-etynyylibentseeni ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

L19082.03 L19082.MD
L19082.03EA 176 EUR
L19082.03 L19082.MD
1-Bromi-4-etynyylibentseeni ≥97%
1-Bromi-4-etynyylibentseeni

Purity: 97% CAS No.: 766-96-1 Molecular Formula: C8H5Br Formula Weight: 181.04 Odour: Odourless Melting Point: 64-67° Boiling Point: 88-90°/16mm Beilstein Reference: 1209726 MDL No.: MFCD00168822

Kaava: C₈H₅Br
Kiehumispiste: 88…90 °C (16 mmHg)
Sulamispiste: 64…67 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00168822
CAS-numero: 766-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR