(R)-(-)-4-Metyyli-2-pentanoli ≥99%

Toimittaja: Thermo Fisher Scientific
Varoitus

L18881.03 L18881.MD
L18881.03EA 290 EUR
L18881.03 L18881.MD
(R)-(-)-4-Metyyli-2-pentanoli ≥99%
(R)-(-)-4-Metyyli-2-pentanoli

Purity: 99% CAS No.: 16404-54-9 Molecular Formula: C6H14O Formula Weight: 102.18 Flash Point: 41°(105°F) Refractive Index: 1.4110 Odour: Slightly sweetish Boiling Point: 130-131° Beilstein Reference: 1718992 Optical Rotation: -21° (neat) UN No.: 2053 MDL No.: MFCD03093077

Kaava: C₆H₁₄O
Kiehumispiste: 130…131 °C
Tiheys: 0,802
Leimahduspiste: 41 °C (105 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03093077
CAS-numero: 16404-54-9
UN: 2053
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR