3,4,5-Trifluorifenyyliboronihappo ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

L18519.03 L18519.MD
L18519.03EA 380 EUR
L18519.03 L18519.MD
3,4,5-Trifluorifenyyliboronihappo ≥97%
3,4,5-Trifluorifenyyliboronihappo

Purity: 97% CAS No.: 143418-49-9 Molecular Formula: C6H4BF3O2 Formula Weight: 175.90 Melting Point: 275-279° Beilstein Reference: 7371914

Kaava: C₆H₄BF₃O₂
Sulamispiste: 275…279 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02093069
CAS-numero: 143418-49-9
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR