1-Boc-3-hydroxypiperidine ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

L17600.03 L17600.06
L17600.03EA 86.6 EUR
L17600.03 L17600.06
1-Boc-3-hydroxypiperidine ≥97%
1-Boc-3-hydroxypiperidine

Purity: 97% CAS No.: 85275-45-2 Molecular Formula: C10H19NO3 Formula Weight: 201.27 Melting Point: 66-70° Beilstein Reference: 4180859

Kaava: C₁₀H₁₉NO₃
Sulamispiste: 66…70 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02093938
CAS-numero: 85275-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR