6-Metyyli-indoli ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

L17509.03 L17509.MD
L17509.03EA 206 EUR
L17509.03 L17509.MD
6-Metyyli-indoli ≥98%
6-Metyyli-indoli

Purity: 98% CAS No.: 3420-02-8 Molecular Formula: C9H9N Formula Weight: 131.18 Flash Point: 110°(230°F) Refractive Index: 1.6071 Melting Point: 29-32° Boiling Point: 112°/5mm Beilstein Reference: 109708 MDL No.: MFCD00005682

Kaava: C₉H₉N
Kiehumispiste: 112 °C (5 mmHg)
Sulamispiste: 29…32 °C
Tiheys: 1,059
Leimahduspiste: >110 °C (230 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005682
CAS-numero: 3420-02-8
UN: 3334
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR