5-jodi-1,2,3-trimetoksibentseeni ≥98%

Toimittaja: Thermo Fisher Scientific

L17414.03 L17414.06
L17414.03EA 160 EUR
L17414.03 L17414.06
5-jodi-1,2,3-trimetoksibentseeni ≥98%
5-jodi-1,2,3-trimetoksibentseeni

Purity: 98+% CAS No.: 25245-29-8 Molecular Formula: C9H11IO3 Formula Weight: 294.09 Melting Point: 85-87° Beilstein Reference: 1529690

Kaava: C₉H₁₁IO₃
Sulamispiste: 85…87 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01318153
CAS-numero: 25245-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR