Silikageeli 99.9%, powder 1,0 µm

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

L16986.14 L16986.22
L16986.14EA 180 EUR
L16986.14 L16986.22
Silikageeli 99.9%, powder 1,0 µm
Silikageeli

Purity: 99.9% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Odour: Odourless pH: 6 Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232

Kaava: SiO₂
MW: 60.08 g/mol
Kiehumispiste: 2230 °C
Sulamispiste: 1710 °C
Tiheys: 2.0
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011232
CAS-numero: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,8493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR