(R)-(+)-α-Ethylbenzylamine ≥99%, ee 98%, ChiPros®

Toimittaja: Thermo Fisher Scientific
Vaara

L16319.06 L16319.14 L16319.03
L16319.06EA 144 EUR
L16319.06 L16319.14 L16319.03
(R)-(+)-α-Ethylbenzylamine ≥99%, ee 98%, ChiPros®
(R)-(+)-α-Ethylbenzylamine

Purity: 99+% CAS No.: 3082-64-2 Molecular Formula: C9H13N Formula Weight: 135.21 Flash Point: 77°(170°F) Refractive Index: 1.5200 Odour: Amine-like Melting Point: -69° Boiling Point: 205° Beilstein Reference: 3029904 Optical Rotation: +20° (neat) UN No.: 2735 MDL No.: MFCD00083057

Kaava: C₉H₁₃N
Kiehumispiste: 205 °C
Sulamispiste: –69 °C
Tiheys: 0,930
Leimahduspiste: 77 °C (170 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00083057
CAS-numero: 3082-64-2
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR