4-Iodophenyl isocyanate ≥97%

Toimittaja: Thermo Fisher Scientific
Vaara

L16000.03 L16000.06
L16000.03EA 87.3 EUR
L16000.03 L16000.06
4-Iodophenyl isocyanate ≥97%
4-Iodophenyl isocyanate

Purity: 97% CAS No.: 15845-62-2 Molecular Formula: C7H4INO Formula Weight: 245.02 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Odour: Pungent Melting Point: 43-47° Boiling Point: 84°/5mm Beilstein Reference: 2716151 UN No.: 2811 MDL No.: MFCD00013875

Kaava: C₇H₄INO
Kiehumispiste: 84 °C (5 mmHg)
Sulamispiste: 43…47 °C
Leimahduspiste: >110 °C (230 °F)
Storage Temperature: Jääkaappi
MDL numero: MFCD00013875
CAS-numero: 15845-62-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR