Ethanolamine hydrochloride ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

L14322.22 L14322.36
L14322.22EA 72.4 EUR
L14322.22 L14322.36
Ethanolamine hydrochloride ≥98%
Ethanolamine hydrochloride

Purity: 98+% CAS No.: 2002-24-6 Molecular Formula: HOCH2CH2NH2.HCl Formula Weight: 97.55 Melting Point: 83-86° Beilstein Reference: 3542892 Merck Reference: 14,3727 MDL No.: MFCD00012892

Kaava: C₂H₈ClNO
Sulamispiste: 83…86 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012892
CAS-numero: 2002-24-6
EINECS: 217-900-6
Merck Index: 14,3727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR