Norbornadiene ≥97% stabiloitu

Toimittaja: Thermo Fisher Scientific
Vaara


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

L13935.ACEA 96.8 EUR
L13935.AC L13935.AE
Norbornadiene ≥97% stabiloitu
Norbornadiene

Stabilized with 250 ppm BHT


Purity: 97% CAS No.: 121-46-0 Molecular Formula: C7H8 Formula Weight: 92.14 Flash Point: -11°(12°F) Refractive Index: 1.4700 Melting Point: -20° Boiling Point: 88-90° Beilstein Reference: 506224 UN No.: 2251 MDL No.: MFCD00082301

Kaava: C₇H₈
Kiehumispiste: 88…90 °C
Sulamispiste: –20 °C
Tiheys: 0,906
Leimahduspiste: –11 °C (–12 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00082301
CAS-numero: 121-46-0
EINECS: 204-472-0
UN: 2251
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR