(2,4-Dimetoksifenyyli)etikkahappo ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

L13784.03 L13784.06
L13784.03EA 58.9 EUR
L13784.03 L13784.06
(2,4-Dimetoksifenyyli)etikkahappo ≥98%
(2,4-Dimetoksifenyyli)etikkahappo

Purity: 98% CAS No.: 6496-89-5 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 108-112° Beilstein Reference: 2694132

Kaava: (C₆H₃(OCH₃)₂)CH₂COOH
Sulamispiste: 108…112 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00183328
CAS-numero: 6496-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR