(2-Jodifenyyli)asetonitriili 96%

Toimittaja: Thermo Fisher Scientific
Vaara

L12767.03 L12767.06
L12767.03EA 64.8 EUR
L12767.03 L12767.06
(2-Jodifenyyli)asetonitriili 96%
(2-Jodifenyyli)asetonitriili

Purity: 96% CAS No.: 40400-15-5 Molecular Formula: C8H6IN Formula Weight: 243.05 Flash Point: 110°(230°F) Refractive Index: 1.6170 Boiling Point: 139-140°/13mm Beilstein Reference: 2083106 UN No.: 3276 MDL No.: MFCD00040888

Kaava: C₈H₆IN
Kiehumispiste: 139…140 °C (13 mmHg)
Tiheys: 1,750
Leimahduspiste: >110 °C (230 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00040888
CAS-numero: 40400-15-5
UN: 3276
ADR: 6.1,III

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR