Trifluoroacetamidine tech. 85%, Technical Grade

Toimittaja: Thermo Fisher Scientific
Vaara

L12422.06 L12422.14 L12422.22
L12422.06EA 222 EUR
L12422.06 L12422.14 L12422.22
Trifluoroacetamidine tech. 85%, Technical Grade
Trifluoroacetamidine

Purity: 85% CAS No.: 354-37-0 Molecular Formula: C2H3F3N2 Formula Weight: 112.05 Storage Temperature: -30°C to -10°C Refractive Index: 1.3820 Boiling Point: 35-37°/11mm Beilstein Reference: 605978 UN No.: 3267 MDL No.: MFCD00041545

Kaava: C₂H₃F₃N₂
Kiehumispiste: 35…37 °C (11 mmHg)
Tiheys: 1,45
Storage Temperature: Pakastin
MDL numero: MFCD00041545
CAS-numero: 354-37-0
UN: 3267
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR