1-Naftyyli-isotiosyanaatti ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

L12418.09
L12418.09EA 323 EUR
L12418.09
1-Naftyyli-isotiosyanaatti ≥98%
1-Naftyyli-isotiosyanaatti

Purity: 98% CAS No.: 551-06-4 Molecular Formula: C11H7NS Formula Weight: 185.25 Odour: Irritating Melting Point: 54-57° Beilstein Reference: 637868 Merck Reference: 14,6411 UN No.: 2923 MDL No.: MFCD00003882

Kaava: C₁₁H₇NS
Kiehumispiste: 100 °C (0,2 mmHg)
Sulamispiste: 54…57 °C
Tiheys: 1,81 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003882
CAS-numero: 551-06-4
EINECS: 208-990-8
UN: 2923
ADR: 8,III
Merck Index: 14,6411

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR