o-Tolyyli-isosyanaatti ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

L12247.06 L12247.14
L12247.06EA 86.6 EUR
L12247.06 L12247.14
o-Tolyyli-isosyanaatti ≥98%
o-Tolyyli-isosyanaatti

Purity: 98% CAS No.: 614-68-6 Molecular Formula: C8H7NO Formula Weight: 133.15 Storage Temperature: 2°C to 8°C Flash Point: 92°(197°F) Refractive Index: 1.5345 Odour: Pungent Boiling Point: 186° Beilstein Reference: 507985 UN No.: 2206 MDL No.: MFCD00002009

Kaava: C₈H₇NO
Kiehumispiste: 186 °C
Tiheys: 1,074
Leimahduspiste: 92 °C (197 °F)
MDL numero: MFCD00002009
CAS-numero: 614-68-6
EINECS: 210-389-0
UN: 2206
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR