Mono-n-dodecyl phosphate 90%, tech.

Toimittaja: Thermo Fisher Scientific
Varoitus

L11941.06 L11941.14
L11941.06EA 290 EUR
L11941.06 L11941.14
Mono-n-dodecyl phosphate 90%, tech.
Mono-n-dodecyl phosphate

Purity: 90% CAS No.: 2627-35-2 Molecular Formula: C12H27O4P Formula Weight: 266.32 Odour: Odourless Melting Point: 39-45° Beilstein Reference: 1710518 MDL No.: MFCD00013946

Kaava: C₁₂H₂₇O₄P
Sulamispiste: 39…45 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013946
CAS-numero: 2627-35-2
EINECS: 220-095-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR