(S)-(+)-2-Amino-3-metyyli-1-butanoli ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

L11300.03 L11300.06 L11300.14
L11300.03EA 74.5 EUR
L11300.03 L11300.06 L11300.14
(S)-(+)-2-Amino-3-metyyli-1-butanoli ≥97%
(S)-(+)-2-Amino-3-metyyli-1-butanoli

Purity: 97% CAS No.: 2026-48-4 Molecular Formula: C5H13NO Formula Weight: 103.17 Storage Temperature: 2°C to 8°C Flash Point: 78°(172°F) Refractive Index: 1.4548 Melting Point: 30-35° Boiling Point: 80-81°/8mm Beilstein Reference: 1719137 Optical Rotation: +17° (c=10 in ethanol) MDL No.: MFCD00064296

Kaava: C₅H₁₃NO
Kiehumispiste: 80…81 °C (8 mmHg)
Sulamispiste: 30…35 °C
Tiheys: 0,926
Leimahduspiste: 78 °C (172 °F)
MDL numero: MFCD00064296
CAS-numero: 2026-48-4
EINECS: 217-975-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR