Norbornene ≥99%

Toimittaja: Thermo Fisher Scientific
Vaara

L11103.22 L11103.36
L11103.22EA 64.6 EUR
L11103.22 L11103.36
Norbornene ≥99%
Norbornene

Purity: 99% CAS No.: 498-66-8 Molecular Formula: C7H10 Formula Weight: 94.16 Flash Point: -15°(5°F) Odour: Pungent Boiling Point: 95-96° Beilstein Reference: 1098544 UN No.: 1325 MDL No.: MFCD00082304

Kaava: C₇H₁₀
Kiehumispiste: 95…96 °C
Sulamispiste: 44…47 °C
Tiheys: 0,955
Leimahduspiste: –15 °C (5 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00082304
CAS-numero: 498-66-8
EINECS: 207-866-0
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR