Diethylene glycol diacrylate ≥75% stabiloitu, tech.

Toimittaja: Thermo Fisher Scientific
Vaara

L10682.14 L10682.22
L10682.14EA 64.8 EUR
L10682.14 L10682.22
Diethylene glycol diacrylate ≥75% stabiloitu, tech.
Diethylene glycol diacrylate

Purity: 75% CAS No.: 4074-88-8 Molecular Formula: C10H14O5 Formula Weight: 214.22 Flash Point: 110°(230°F) Refractive Index: 1.4620 Boiling Point: 94°/0.2mm Beilstein Reference: 1953711 UN No.: UN2810 MDL No.: MFCD00014939

Kaava: C₁₀H₁₄O₅
Kiehumispiste: 94 °C (0,2 mmHg)
Tiheys: 1,118
Leimahduspiste: >110 °C (230 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00014939
CAS-numero: 4074-88-8
EINECS: 223-791-6
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR