2,6-Diisopropylphenyl isocyanate ≥94%

Toimittaja: Thermo Fisher Scientific
Vaara

L10613.06 L10613.14 L10613.03
L10613.06EA 179 EUR
L10613.06 L10613.14 L10613.03
2,6-Diisopropylphenyl isocyanate ≥94%
2,6-Diisopropylphenyl isocyanate

Purity: 94% CAS No.: 28178-42-9 Molecular Formula: C13H17NO Formula Weight: 203.29 Storage Temperature: 2°C to 8°C Flash Point: 107°(224°F) Refractive Index: 1.5180 Boiling Point: 116-117°/10mm Beilstein Reference: 2645721 UN No.: 2206 MDL No.: MFCD00008882

Kaava: C₁₃H₁₇NO
Kiehumispiste: 116…117 °C (10 mmHg)
Tiheys: 0,951
Leimahduspiste: 107 °C (224 °F)
MDL numero: MFCD00008882
CAS-numero: 28178-42-9
EINECS: 248-885-4
UN: 2206
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR