Ethyl-2-piperidinecarboxylate ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

L10532.03 L10532.06 L10532.14
L10532.03EA 49.9 EUR
L10532.03 L10532.06 L10532.14
Ethyl-2-piperidinecarboxylate ≥98%
Ethyl-2-piperidinecarboxylate

Purity: 98+% CAS No.: 15862-72-3 Molecular Formula: C8H15NO2 Formula Weight: 157.21 Flash Point: 46°(114°F) Refractive Index: 1.4560 Boiling Point: 216-217° Beilstein Reference: 81694 UN No.: 1993 MDL No.: MFCD00005980

Kaava: C₈H₁₅NO₂
Kiehumispiste: 216…217 °C
Tiheys: 1,006
Leimahduspiste: 46 °C (114 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005980
CAS-numero: 15862-72-3
EINECS: 239-990-6
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR