cis-4-Cyclohexene-1,2-dicarboxylic acid ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

L09392.14 L09392.22
L09392.14EA 90.4 EUR
L09392.14 L09392.22
cis-4-Cyclohexene-1,2-dicarboxylic acid ≥98%
cis-4-Cyclohexene-1,2-dicarboxylic acid

Purity: 98% CAS No.: 2305-26-2 Molecular Formula: C8H10O4 Formula Weight: 170.17 Melting Point: 165-170° Beilstein Reference: 2050419

Kaava: C₈H₁₀O₄
Sulamispiste: 165…170 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00070482
CAS-numero: 2305-26-2
EINECS: 218-974-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR