6-Methylnicotinic acid ≥99%

Toimittaja: Thermo Fisher Scientific
Varoitus

L08877.03 L08877.06 L08877.14
L08877.03EA 67.9 EUR
L08877.03 L08877.06 L08877.14
6-Methylnicotinic acid ≥99%
6-Methylnicotinic acid

Purity: 99% CAS No.: 3222-47-7 Molecular Formula: C7H7NO2 Formula Weight: 137.14 Melting Point: 210-215° Beilstein Reference: 112049

Kaava: C₇H₇NO₂
Sulamispiste: 210…215 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006341
CAS-numero: 3222-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR