Dibenzyl phosphate ≥98%
Toimittaja: Thermo Fisher Scientific
L08813.03 L08813.06 L08813.14
L08813.03EA 60.8 EUR
L08813.03 L08813.06 L08813.14
Dibenzyl phosphate ≥98%
Dibenzyl phosphate
Purity: 98% CAS No.: 1623-08-1 Molecular Formula: C14H15O4P Formula Weight: 278.24 Storage Temperature: 2°C to 8°C Melting Point: 78-81° Beilstein Reference: 2055755 MDL No.: MFCD00004775