Aleuritic acid 95%

Toimittaja: Thermo Fisher Scientific

L08542.06 L08542.14
L08542.06EA 122 EUR
L08542.06 L08542.14
Aleuritic acid 95%
Aleuritic acid

Purity: 95% CAS No.: 533-87-9 Molecular Formula: C16H32O5 Formula Weight: 304.43 Odour: Odourless Melting Point: 97-101° Beilstein Reference: 1727724 MDL No.: MFCD00135596

Kaava: C₁₆H₃₂O₅
Sulamispiste: 97…101 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00135596
CAS-numero: 533-87-9
EINECS: 208-578-8
Merck Index: 14,231

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR